Learn More
Thermo Scientific™ Sterilin™ Round Base Soda Glass Tubes
Product Code. 12872863
Change view
Click to view available options
Capacity (Metric):
3 mL
5 mL
Diameter (Metric):
10 mm
12 mm
Height (Metric):
75 mm
No. per Case:
1000
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Height (Metric) No. per Case unitSize
12872863 3 mL 10 mm 75 mm 1000 Pack of 1000
12882863 5 mL 12 mm 75 mm 1000 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12872863 Supplier Thermo Scientific™ Supplier No. 49684
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rely on these round base glass tubes for excellent chemical resistance for samples to 2mL or 4mL. Test tube profile for streamlined handling and excellent mixing capability.
Rely on these round base glass tubes for excellent chemical resistance for samples to 2mL or 4mL. Thermo Scientific™ Sterilin™ Round Base Soda Glass Tube profile enables streamlined handling and excellent mixing capability.
NOT AVAILABLE IN AMERICA
  • Provides greater chemical resistance than plastic
  • Test tube profile with rim
  • Supplied without caps
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 10 mm
Product Type Soda Glass Tube
Capacity (Metric) 3 mL
Material Soda Glass
No. per Case 1000
No. per Pack 250
Sterility Sterile
Packaging Quantity 1000 Case (250 Pack)
Capacity (English) 0.101 fl oz.
Height (Metric) 75 mm