Learn More
Thermo Scientific™ Sterilin™ Round Base Soda Glass Tubes
Description
Rely on these round base glass tubes for excellent chemical resistance for samples to 2mL or 4mL. Thermo Scientific™ Sterilin™ Round Base Soda Glass Tube profile enables streamlined handling and excellent mixing capability.
NOT AVAILABLE IN AMERICA
- Provides greater chemical resistance than plastic
- Test tube profile with rim
- Supplied without caps
Ordering Information
- Not available in all geographic regions
Specifications
Specifications
| Diameter (Metric) | 12 mm |
| Product Type | Soda Glass Tube |
| Capacity (Metric) | 5 mL |
| Material | Soda Glass |
| No. per Case | 1000 |
| No. per Pack | 250 |
| Sterility | Sterile |
| Packaging Quantity | 1000 Case (250 Pack) |
| Capacity (English) | 0.169 fl oz. |
| Height (Metric) | 75 mm |