Learn More
Sampling Systems™ SteriWare™ Disposable Pallet Knife
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
260 mm
300 mm
Length (Metric) Blade:
150 mm
200 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Descrizione
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifica
Specifica
| Description | Individually Bagged |
| Sterility | Sterile |
| Color | White |
| Length (Metric) Blade | 200 mm |
| Width (Metric) | 45 mm |
| Material | HDPE |
| Length (Metric) | 300 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto