Learn More
Sampling Systems™ SteriWare™ Disposable Pallet Knife
BIOPHARMA
Product Code. 15750678 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
260 mm
300 mm
Length (Metric) Blade:
150 mm
200 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 15750678
Brand: Sampling Systems™ 8175H150
Only null left
Add to basket
Add to basket
Constructed of food/pharma grade HDPE, making it ideal for manufacturing areas. Sampling Systems™ SteriWare™ Disposable Pallet Knife is good for hygienic applications and requires no cleaning.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Description Individually Bagged
Sterility Non-sterile
Color White
Length (Metric) Blade 150 mm
Width (Metric) 40 mm
Material HDPE
Length (Metric) 260 mm