Learn More
Sampling Systems™ SteriWare™ Funnels
BIOPHARMA
Product Code. 17594656 Shop All Sampling Systems Products
Click to view available options
Diameter (Metric)Top:
50 mm
Unit Size:
Pack of 100
Product Code. 17594656
Brand: Sampling Systems™ 8370H50
Only null left
Add to basket
Add to basket
SteriWare™ Funnels are part of a large range of high quality single use liquid funnels. A large range of sizes in sterile and non sterile mean that we will have the right item for you. Simply unwrap and use.
  • Disposable, single use, sterile liquid funnels.
  • They are ideal for use in laboratories and in manufacturing areas.
  • These funnels are made from food grade polymers and are designed to be used straight from the bag.
  • SteriWare™ Funnels are available sterile and non-sterile.
  • Simply unwrap and use.
  • They are ideal for dispensing liquids and creams.
  • Each funnel has a small lug on the rim so you can hold the funnel and not get your fingers wet.
  • Made from high quality HDPE which conforms to the latest FDA and EU food requirements.
  • BSE free.
  • Cleanroom manufactured.
  • Available sterile (SAL 10-6) and non-sterile.
  • 50 mm, 100 mm and 200 mm top diameters.
  • All orders are supplied with certificates of conformity and sterilisation where applicable.
Specifications
Diameter (Metric)Top 50 mm
Material HDPE