Learn More
Sampling Systems™ SteriWare™ LiquiDip Mono
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
225 mL
Unit Size:
Pack of 20
Description
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Suitable for taking cross sectional samples
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Available in 2 different lengths
- Push the sampler into the product, ensuring the sampler is in the open position
- Push the sampler against the base of the container to close the valve
- Remove the sampler and pour out the liquid into a suitable container
- No cross contamination
- No cleaning required
Specifications
Specifications
| Product Type | Liquid Sampler |
| Material | HDPE |
| Includes | Valve at one end |
| Capacity (Metric) | 225 mL |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Diameter (Metric) Outer | 21 mm |
| Length (Metric) | 1100 mm |