Learn More
Sterol carrier protein 2 Antibody (OTI1E4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74401C
Product Code. 18167666
  • € 751.00 / 0.10mL
Description
Sterol carrier protein 2 Monoclonal specifically detects Sterol carrier protein 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterol carrier protein 2
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
SCP2
Full length recombinant protein of human Sterol carrier protein 2 (NP_002970) produced in HEK293T cells.
0.1 mL
6342.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI1E4
Western Blot, Immunohistochemistry
DKFZp686C12188, DKFZp686D11188, NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, sterol carrier protein 2EC 2.3.1.176, Sterol carrier protein X
Mouse
Affinity Purified
Primary
Human, Mouse, Rat