Learn More
STI1 Antibody (3E4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26299-100ul
 View more versions of this product
Product Code. 30024442
  • € 533.00 / 100µL
Description
STI1 Monoclonal antibody specifically detects STI1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STI1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
3E4
Western Blot 1:500-1:5000, ELISA
Hop, HOPSTI1L, Hsc70/Hsp90-organizing protein, Hsp70/Hsp90-organizing protein, IEF-SSP-3521, NY-REN-11 antigen, Renal carcinoma antigen NY-REN-11, STI1P60, stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521
A synthesized peptide derived from Human STI1 [UniProt P31948]
100 μL
Core ESC Like Genes, Stem Cell Markers
10963
Store at -20 to -70C. Avoid freeze-thaw cycles.