missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STIM1 Antibody (CDN3H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-55300
This item is not returnable.
View return policy
Description
STIM1 Monoclonal specifically detects STIM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| STIM1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 5 ug/ml | |
| GOKD11S4896E, stromal interaction molecule 1 | |
| Mouse | |
| 84 kDa | |
| 0.1 mg | |
| Cancer, Cell Cycle and Replication | |
| 6786 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CDN3H4 | |
| Unconjugated | |
| Q13586 | |
| STIM1 | |
| Synthetized peptide from C-terminal cytoplasmic part of STIM1. (NM_003156.3). | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody CDN3H4 reacts with human and rodent STIM1, a 84 kDa essential and conserved regulator of store-operated Ca2+ channel function. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction