missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STIM1 Antibody (CDN3H4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-55300H
This item is not returnable.
View return policy
Description
STIM1 Monoclonal antibody specifically detects STIM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| STIM1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CDN3H4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| GOKD11S4896E, stromal interaction molecule 1 | |
| Synthetized peptide from C-terminal cytoplasmic part of STIM1. (NM_003156.3). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction | |
| 6786 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction