missing translation for 'onlineSavingsMsg'
Learn More

STIM1 Mouse anti-Human, Mouse, Rat, Clone: 1D3A3, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen STIM1
Clone 1D3A3
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887835
View Documents
Proteintech
66189-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16877835
View Documents
Proteintech
66189-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a type 1 transmembrane protein that mediates Ca2+ influx after depletion of intracellular Ca2+ stores by gating of store-operated Ca2+ influx channels (SOCs). It is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocrotical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in maligcies and disease that involve this region, as well as early hematopoiesis, by mediating attachment to stromal cells. This gene is oriented in a head-to-tail configuration with the ribonucleotide reductase 1 gene (RRM1), with the 3′ end of this gene situated 1.6 kb from the 5′ end of the RRM1 gene.
Specifications

Specifications

STIM1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
20866, 361618, 6786
STIM1 Fusion Protein Ag17914
Primary
-20°C
STIM1
1D3A3
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Unconjugated
Mouse
Mouse, Rat, Human
P70302, P84903, Q13586
Stim1
IgG2a
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.