missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody (OTI4H1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74402B
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| STING/TMEM173 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TMEM173 | |
| Full length recombinant protein of human STING/TMEM173 (NP_938023) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI4H1 | |
| Western Blot, Immunohistochemistry | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Immunology, Innate Immunity | |
| 340061.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction