missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | STK19 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227300
|
Novus Biologicals
NBP3-35338-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230047
|
Novus Biologicals
NBP3-35338-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STK19 Polyclonal antibody specifically detects STK19 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| STK19 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 8859 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D6S60, D6S60E, EC 2.7.11.1, G11nuclear serine/threonine protein kinase, HLA-RP1, MGC117388, MHC class III HLA-RP1, Protein G11, Protein RP1, RP1serine/threonine-protein kinase 19, serine/threonine kinase 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 75-364 of human STK19 (NP_004188.1).,, Sequence:, GTPGETVRHCSAPEDPIFRFSSLHSYPFPGTIKSRDMSWKRHHLIPETFGVKRRRKRGPVESDPLRGEPGSARAAVSELMQLFPRGLFEDALPPIVLRSQVYSLVPDRTVADRQLKELQEQGEIRIVQLGFDLDAHGIIFTEDYRTRVLKACDGRPYAGAVQKFLASVLPACGDLSFQQDQMTQTFGFRDSEITHLVNAGVLTVRDAGSWWLAVPGAGRFIKYFVKGRQAVLSMVRKAKYRELLLSELLGRRAPVVVRLGLTYHVHDLIGAQLVDCISTTSGTLLRLPET | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title