Learn More
Invitrogen™ Human STK22D (TSSK1), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length
Specifications
Specifications
| Accession Number | Q9BXA7 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.05% NP-40, 0.5 mM EDTA, 100 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 83942 |
| Molecular Weight (g/mol) | 46.7 kDa |
| Name | Human STK22D (TSSK1), His Tag |
| Quantity | 10 μg |
| Regulatory Status | For research use only. Not for use in diagnostic procedures |
| Gene Alias | FKSG81; serine/threonine kinase 22 A (spermiogenesis associated); serine/threonine kinase 22 D (spermiogenesis associated); serine/threonine kinase FKSG81; serine/threonine-protein kinase 22 A; spermiogenesis associated; spermiogenesis associated 4; SPOGA1; SPOGA4; Stk22a; STK22D; testis specific serine kinase 1 B; testis tissue sperm-binding protein Li 55 e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1 B; testis-specific serine/threonine kinase; testis-specific serine/threonine-protein kinase 1; TSK1; TSK-1; Tssk; TSSK1; TSSK-1; TSSK1B |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.