missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569552
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
STK25 (YSK1) is a serine/threonine kinase that belongs to the GCK family of kinases. It has been implicated as an upstream regulator of MAP kinase signaling pathways. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
| STK25 | |
| Polyclonal | |
| Unconjugated | |
| STK25 | |
| 1500019J11Rik; AU018434; C86992; hypothetical protein; serine/threonine kinase 25; serine/threonine kinase 25 (STE20 homolog, yeast); serine/threonine kinase 25 (Ste20, yeast homolog); serine/threonine kinase 25 (yeast); serine/threonine-protein kinase 25; SOK1; SOK-1; Ste20, yeast homolog; ste20/oxidant stress response kinase 1; Ste20-like; ste20-like kinase; sterile 20/oxidant stress-response kinase 1; STK25; STK25 protein; Yeast Sps1/Ste20-related kinase 1; Ysk1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 373542 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| STK25 | |
| synthetic peptide corresponding to a region of rat STK25. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction