missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32A Polyclonal Antibody

STK32A Polyclonal Antibody
Brand: Invitrogen PA598765
This item is not returnable.
View return policy
Description
STK32A Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| STK32A | |
| Polyclonal | |
| Unconjugated | |
| STK32A | |
| A930015B13Rik; CTB-108O6.2; serine/threonine kinase 32A; serine/threonine-protein kinase 32A; Stk32a; YANK1; Yet another novel kinase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 202374 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WU08 | |
| STK32A | |
| Recombinant Human Serine/threonine-protein kinase 32A protein (1-166AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction