missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STK33 Recombinant Rabbit Monoclonal Antibody (1E4W7)
GREENER_CHOICE

Product Code. 17246883 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17246883 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17246883 Supplier Invitrogen™ Supplier No. MA538008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: SH-SY5Y Immunogen sequence: KEWKNNPESV EENTTEEKNK PSTEEKLKSY QPWGNVPDAN YTSDEEEEKQ STAYEKQFPA TSKDNFDMCS SSFTSSKLLP AEIKGEMEKT PVTPSQGTAT KYPAKSGALS RTKKKL.

STK33 is a serine/threonine kinase involved in signal transduction and cellular communication. STK33 gene ontology annotations related to this gene include ATP binding; protein kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK33
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1E4W7
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STK33
Gene Accession No. Q924X7, Q9BYT3
Gene Alias 4921505G21Rik; AW061264; serine/threonine kinase 33; serine/threonine-protein kinase 33; Stk33
Gene Symbols STK33
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 399-514 of human STK33 (Q9BYT3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117229, 65975
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.