missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528897
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D, IMR32, Jurkat, HeLa, mouse brain. Predicted reactivity: Mouse (97%), Rat (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.
Specifications
| STK36 | |
| Polyclonal | |
| Unconjugated | |
| STK36 | |
| 1700112N14Rik; B930045J24; FU; Fused; Fused homolog; Kiaa1278; mKIAA1278; serine/threonine kinase 36; serine/threonine kinase 36 (fused homolog, Drosophila); serine/threonine-protein kinase 36; STK36; testicular tissue protein Li 188 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 269209, 27148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q69ZM6, Q9NRP7 | |
| STK36 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human STK36. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction