missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK38L Antibody (OTI4D8), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74403AF488
This item is not returnable.
View return policy
Description
STK38L Monoclonal specifically detects STK38L in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| STK38L | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| STK38L | |
| Recombinant protein fragment corresponding to amino acids 1-281 of human STK38L (NP_055815) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4D8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.11, EC 2.7.11.1, KIAA0965nuclear Dbf2-related 2, NDR2 protein kinase, NDR2serine/threonine-protein kinase 38-like, Nuclear Dbf2-related kinase 2, serine/threonine kinase 38 like | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 23012.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction