missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK39 Antibody (OTI4E3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74406PEATT594
This item is not returnable.
View return policy
Description
STK39 Monoclonal specifically detects STK39 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| STK39 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| STK39 | |
| Full length recombinant protein of human STK39 (NP_037365) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI4E3 | |
| Flow Cytometry | |
| DCHTsmall intestine SPAK-like kinase, DKFZp686K05124, EC 2.7.11, EC 2.7.11.1, PASK, proline-alanine-rich STE20-related kinase, serine threonine kinase 39, Serine/threonine-protein kinase 39, SPAKserine threonine kinase 39 (STE20/SPS1 homolog, yeast), STE20/SPS1 homolog, STE20/SPS1-related proline-alanine-rich protein kinase, Ste20-like protein kinase, Ste-20-related kinase | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 27347.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction