missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK4 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-39128C
This item is not returnable.
View return policy
Description
STK4 Polyclonal antibody specifically detects STK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| STK4 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1 | |
| The immunogen recognized by this antibody maps to the C-terminus of human Mammalian Sterile 20-like 1 (Serine/Threonine Kinase 4) using the numbering given in Swiss Prot entry Q13043 (GeneID 6789). | |
| 0.1 mL | |
| Apoptosis, Phospho Specific | |
| 6789 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction