missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99813-50ul
This item is not returnable.
View return policy
Description
STK4 Polyclonal antibody specifically detects STK4 in Human samples. It is validated for ELISA
Specifications
| STK4 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1 | |
| Produced in rabbits immunized with purified, recombinant Human STK4 (Uniprot#: Q13043-1; Glu2-Phe487) | |
| 50 μg | |
| Apoptosis | |
| 6789 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction