missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STMN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10586-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the stathmin family of phosphoproteins. Stathmin proteins function in microtubule dynamics and signal transduction. The encoded protein plays a regulatory role in neuronal growth and is also thought to be involved in osteogenesis. Reductions in the expression of this gene have been associated with Down′s syndrome and Alzheimer′s disease. Alternatively spliced transcript variants have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 6.Specifications
| STMN2 | |
| Polyclonal | |
| Unconjugated | |
| STMN2 | |
| Protein SCG10, SCG10, SCGN10, SGC10, Stathmin 2, stathmin like 2, STMN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11075, 20257, 84510 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21818, P55821, Q93045 | |
| Stmn2 | |
| STMN2 Fusion Protein Ag0929 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction