Learn More
STMN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12027-1-AP-20UL
 View more versions of this product
Product Code. 16879493
  • € 145.00 / 20µL
Description
RB3 (Stathmin-4), encoded by STMN4 gene, is a neuron-specific homologue of the SCG10/stathmin family proteins, possessing a unique N-terminal membrane-associated domain and the stathmin-like domain at the C terminus, which promotes microtubule (MT) catastrophe and/or tubulin sequestering. Stathmin is a ubiquitous cytosolic phosphoprotein, preferentially expressed in the nervous system, and described as a relay integrating diverse intracellular signaling pathways. RB3 is consistently up-regulated in differentiated neuroblastoma cells, and stable expression of transfected RB3 could induce differentiation, suggesting a role of RB3 during neuroblastoma differentiation. Moreover, increased RB3 is a unique response protein in the stathmin-related proteins following optical nerve axotomy and RB3 may be a critical target for neurodegenerative diseases.
Specifications
Specifications
STMN4
Polyclonal
Unconjugated
STMN4
RB3, Stathmin 4, stathmin like 4, Stathmin like protein B3, STMN4
Rabbit
Antigen Affinity Chromatography
RUO
56471, 79423, 81551
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63042, P63043, Q9H169
Stmn4
STMN4 Fusion Protein Ag2657
20 μL
Primary
Human, Rat, Mouse