missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STOM Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110443

Product Code. 17118373

  • € 493.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KIIKEYERAI IFRLGRILQG GAKGPGLFFI LPCTDSFIKV DMRTISFDIP PQEILTKDSV TISVDGVVYY RVQNATLAVA NITNADSATR LLAQTTLRNV LGTKNLSQIL SDREEIAHNM QSTLDDATDA WGIKVERVEI KDVKLPVQLQ RAMAAEAEAS REARAKVIAA EGEMNASRAL KEASMVITES PAALQLRYLQ TLTTIAAEKN STIVFPLPID MLQGIIGAKH SHLG.

Thought to regulate cation conductance. May regulate ACCN1 and ACCN3 gating.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STOM
Polyclonal
Unconjugated
STOM
BND7; EPB7; Epb7.2; EPB72; erythrocyte band 7 integral membrane protein; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte protein band 7.2; erythrocyte protein band 7.2; protein 7.2b; erythrocyte surface protein band 7.2; protein 7.2b; STOM; Stomatin
Rabbit
Affinity Chromatography
RUO
13830, 2040, 296655
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27105, P54116
STOM
Recombinant fusion protein containing a sequence corresponding to amino acids 55-288 of human STOM (NP_004090.4).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.