missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stomatin Antibody (33F5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42660JF646
This item is not returnable.
View return policy
Description
Stomatin Monoclonal antibody specifically detects Stomatin in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Stomatin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| BND7EPB7, EPB72erythrocyte band 7 integral membrane protein, erythrocyte membrane protein band 7.2 (stomatin), Protein 7.2b, Stomatin | |
| Recombinant human Stomatin (55-288aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience | |
| 2040 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunofluorescence | |
| 33F5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction