Learn More
STOML2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Product Code. 18050336 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050336 20 μL 20µL
18018085 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18050336 Supplier Proteintech Supplier No. CL4881034820UL
Rabbit Polyclonal Antibody
The STOML2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human STOML2. This antibody recognizes human, mouse, rat antigen. The STOML2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Antigen STOML2
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols EPB72 like protein 2, HSPC108, SLP 2, SLP2, stomatin (EPB72) like 2, Stomatin like protein 2, STOML2
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 30968
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG