Learn More
Lenz Laborglasintrumente Stopcocks, Straight Bore, with PTFE Plug
Product Code. p-7222549
Click to view available options
Plug Bore:
1.5 mm
4 mm
6 mm
Unit Size:
Each
Pack of 10
Product Code. 10777561
Brand: Lenz Laborglasintrumente 02161201
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12541-1
  • With PTFE plug and screwthread retaining device
  • One straight bore
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Plug Bore 1.5 mm
Material Borosilicate Glass (3.3)