missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STP 10 Model Buffer Solution pH 10, WTW™
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
Technical buffering solutions for pH 2.00, pH 4.01, pH 7.00 and pH 10.01: pH 4.01, pH 7.00. This allows to adapt the bulk sizes easily to individual use. The buffer 10.0 is a special case:It is only still suitable for inoLab™ device of the series level 1, 2, 3, portable meters 3xx as well as for various other old devices. All modern technical buffering solutions are traceable to PTB (Physical Technical Government Agency) or NIST (National Institute of Standards and Technology, USA). Their accuracy is ± 0,03 pH.
Specifications
Specifications
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Packaging | Bottle |
| Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction