Learn More
Model Buffer Solutions, WTW™
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
- Technical buffering solutions for pH 4.01
- Available in the practical 50 ml bottles including dosing device
- All modern technical buffering solutions are traceable to NIST (National Institute of Standards and Technology, USA)
- Their accuracy is ± 0,03 pH
- Every bottle has its own lot reference with the expiration date
Specifications
Specifications
| pH | 4 |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Packaging | Bottle |
| Traceability to NIST | Yes |