missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STPG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113245
This item is not returnable.
View return policy
Description
STPG2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
STPG2 (Sperm-tail PG-rich repeat-containing protein 2) associated diseases include Complex Chromosomal Rearrangement and Cone-Rod Dystrophy 18. An important paralog of this gene is STPG1.
Specifications
| STPG2 | |
| Polyclonal | |
| Unconjugated | |
| STPG2 | |
| C4orf37; sperm tail PG-rich repeat containing 2; sperm-tail PG-rich repeat containing 2; sperm-tail PG-rich repeat-containing protein 2; STPG2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 285555 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N412 | |
| STPG2 | |
| Recombinant Human Sperm-tail PG-rich repeat-containing protein 2 protein (201-459AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction