missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA6 Antibody (496613), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5904U
This item is not returnable.
View return policy
Description
STRA6 Monoclonal antibody specifically detects STRA6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| STRA6 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64220 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 496613 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| FLJ12541, MCOPS9, stimulated by retinoic acid gene 6 homolog (mouse), stimulated by retinoic acid gene 6 protein homolog | |
| NS0 mouse myeloma cell line transfected with human STRA6, Met1-Pro667, Accession # AAH25256 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction