missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA8 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98692UV
This item is not returnable.
View return policy
Description
STRA8 Polyclonal antibody specifically detects STRA8 in Human samples. It is validated for Western Blot
Specifications
| STRA8 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 346673 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| stimulated by retinoic acid gene 8 homolog (mouse), stimulated by retinoic acid gene 8 protein homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human STRA8. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction