missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Straight Conical Connectors

Tapered straight connectors

Brand:  Kartell™ 0051100

Product Code. 15508275

  • ONLINE EXCLUSIVE PRICE
    € 130.00
    € 55.38 / Pack of 100
    Save € 74.62
    57% Off
Only null left
Estimated Shipment: 16-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tapered
  • Crest Outer Diameter 4,5 / 7 / 8,2mm
  • Nominal Outer Diameter 6 / 7 / 8mm
  • Bore 3,5mm
  • Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Promotions

    Promotions

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.