Learn More
Lenz Laborglasintrumente™ Straight Receiver Adapters
Click to view available options
Thread Size:
GL 18
Unit Size:
Each
Description
- Receiver adapters acc. to DIN 12594
- With NS socket and NS cone
- With drain tip
- Vacuum connection: thread GL18
- Made of DURAN™ tubing
Specifications
Specifications
| Cone Size | NS 19/26 |
| Material | Duran Tubing |
| Quantity | 1 |
| Standard Taper Joint No. | NS 19/26 |
| Thread Size | GL 18 |