Learn More
Lenz Laborglasintrumente™ Straight Splash Heads acc. to Stutzer
Splash head for separation of drops (direct placement onto distillation flasks)
Brand:  Lenz Laborglasintrumente™ 05035032
Additional Details : Weight : 0.50000kg
Product Code. 10345641
  • € 82.15 / Each
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
  • Straight shape
  • With NS socket and NS cone
  • For separation of drops (direct placement onto distillation flasks)
  • Made of DURAN™ tubing
Specifications
Specifications
DURAN™ tubing
1
Cone - NS 24/29, Socket - NS 19/26