Product Code. 15463635
Click to view available options
Capacity (Metric):
10 mL
15 mL
20 mL
25 mL
35 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Outer:
Outside: 101mm
Outside: 127mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Height (Metric):
18mm
22mm
29mm
34mm
41mm
45mm
59mm
65mm
89mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Capacity (Metric)
Diameter (Metric) Outer
Height (Metric)
Unit Size
15463635
500 mL
Outside: 101mm
22mm
Each
15403595
10 mL
Outside: 27mm
34mm
Each
15463595
15 mL
Outside: 33mm
41mm
Each
15423605
20 mL
Outside: 33mm
41mm
Each
15483605
25 mL
Outside: 45mm
22mm
Each
15403615
35 mL
Outside: 46mm
89mm
Each
15463615
55 mL
Outside: 46mm
22mm
Each
15443625
1000 mL
Outside: 127mm
65mm
Each
15493625
100 mL
Outside: 59mm
18mm
Each
15433635
55 mL
Outside: 47mm
59mm
Each
15483635
250 mL
Outside: 82mm
45mm
Each
15493635
75 mL
Outside: 51mm
29mm
Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15463635
Supplier
Thermo Scientific Alfa Aesar
Supplier No.
036087.KT
Estimated Shipment: 09-02-2027Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Description
Straight Wall
Material
Zirconium
Capacity (Metric)
500 mL
Diameter (Metric) Outer
Outside: 101mm
Height (Metric)
22mm
Product Type
Zirconium Crucible
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?