Learn More
IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity FPLC column
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 mL
5 mL
Unit Size:
1mL
5mL
Description
- Strep-Tactin™ 4Flow™ high capacity FPLC columns are designed for purifying Strep-tag™II and Twin-Strep-tag™ proteins via affinity chromatography in automated protein purification with HPLC/FPLC devices such as Äkta™ systems (Cytiva).
- Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
- 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
- Multiple regeneration cycles possible without loss of binding capacity
- Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
- Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications
Specifications
| Binding Capacity | 20 mg/mL resin |
| Capacity (Metric) | 5 mL |
| Description | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Form | Resin Slurry |
| Quantity | 5 mL |
| Protein Form | Tetrameric protein |
| Target | Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin |
| Type | Column |
| Final Product Type | Affinity chromatography equipment |
| Show More |