Learn More
IBA Lifesciences Strep-Tactin™ protein as lyophilized powder
Strep-Tactin™ protein as lyophilized powder can be used for reversible immobilization of biotinylated analytes or Strep-tag™II and Twin-Strep-tag™ fusion proteins
€ 115.00 - € 327.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17079133
|
IBA Lifesciences
2-1204-001 |
1 mg |
€ 115.00
1mg |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
17171921
|
IBA Lifesciences
2-1204-005 |
5 mg |
€ 327.00
5mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Description
Strep-Tactin™ is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tag™II and Twin-Strep-tag™ fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).Specifications
>50 nmol/mg |