missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strep Tag II Monoclonal Antibody (1810CT579.47.56.10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA537747
Additional Details : Weight : 0.01000kg
Description
The Strep-tag II is an eight-residue minimal peptide sequence (Trp-Ser-His-Pro-Gln-Phe-Glu-Lys) that exhibits intrinsic affinity toward streptavidin and can be fused to recombinant proteins in various fashions. The original Strep-tag was a nine amino acid peptide with high specificity and affinity towards streptavidin which allows the simple purification of protein by use of affinity columns, but required addition to only the C-terminus of recombinant proteins. To also allow a Strep-tag to be placed at the N-terminus of recombinant proteins, it was re-engineered and re-named Strep-tag II.Specifications
Strep Tag II | |
Monoclonal | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 κ |
Western Blot | |
1810CT579.47.56.10 | |
Unconjugated | |
Streptavidin; Streptomyces | |
KLH conjugated synthetic peptide of STREP II TAG | |
200 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |