Learn More
IBA Lifesciences Streptamer™ CD3/CD28 Premix for T Cell Expansion
Tienda IBA Lifesciences Productos
missing translation for 'orderingAttributeHoverText'
:
Capacity: 480 μL
Descripción
- For 1.6 x 107 cells in total
- The vial contains CD3 Fab-Strep, CD28 Fab-Strep and multimeric Strep-Tactin™ for cell expansion, premixed and ready to use, in a total volume of 480 μL
- With this ready-to-use reagent you will benefit from a quick protocol
- The multimers can be removed from the cells by the addition of D-Biotin
- Suspended in sterile PBS
- Product is not classified as hazardous according to (EC) No 1272/2008 [CLP]
- Solution is sterile filtered and does not contain any preservatives
- Please handle under sterile conditions
Especificaciones
Especificaciones
| Capacity (Metric) | 480 μL |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | For stimulation of 1.6 x 107 cells |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido