missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin Antibody (1C2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04345CL1
This item is not returnable.
View return policy
Description
Streptavidin Monoclonal antibody specifically detects Streptavidin in Bacteria samples. It is validated for ELISA,Western Blot
Specifications
| Streptavidin | |
| Monoclonal | |
| CoraFluor 1 | |
| SA V1; SA V2; Streptavidin V1; Streptavidin V2 | |
| Recombinant Streptavidin (25-183aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| ELISA, Western Blot | |
| 1C2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction