missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin-Fluorescein Isothiocyanate (FITC) Conjugate
Click to view available options
Quantity:
50 μL
Unit Size:
Each
Description
Streptavidin-FITC conjugate is typically used in the immunodetection of biotinylated proteins. This conjugate is suitable for use in immunomicroscopy and fluorescence-activated cell sorting and analysis. The amount of conjugate provided is sufficient for 25 immunostaining reactions using 1 ml of working solution per reaction as appropriate.
Specifications
Specifications
| Concentration | 0.75 to 1.2 mg/mL |
| Conjugate | FITC (Fluorescein) |
| Quantity | 50 μL |
| Content And Storage | Store in refrigerator (2—8°C). |
| Product Type | Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction