Learn More
Invitrogen™ Streptavidin, (PE-TR)
Product Code. 10552635
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10552635 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10552635 Supplier Invitrogen™ Supplier No. SA1017
Streptavidin, (PE-TR)
  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 mg/mL
Conjugate PE-Texas Red
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 0.5 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, PE-TR conjugate.
Store in refrigerator (2–8°C).
Excitation/Emission 488/615
Flow Cytometer Laser Lines 488 or 561, 488
Shipping Condition Wet Ice
Form Solution
Product Line Texas Red
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye PE-Texas Red