Learn More
Invitrogen™ eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Specifications
Specifications
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Quantity | 100 μg |
| Form | Liquid |
| Product Line | eBioscience |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Target Molecule | Biotin |
| Label or Dye | PerCP-eFluor 710 |