missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1398-0.5ML
This item is not returnable.
View return policy
Description
Streptococcus B Polyclonal specifically detects Streptococcus B in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Streptococcus B | |
| Polyclonal | |
| Unconjugated | |
| 10 mM PBS (pH 7.2) with 0.1% sodium azide | |
| Rabbit | |
| 0.5 mL | |
| This targets a type specific carbohydrate of group B Streptococcus and does not react with other Strep groups. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunofluorescence | |
| Beta strep, Group b streptococci, Group b streptococcus, Strep b | |
| Streptococci, Group B | |
| Primary | |
| Bacteria | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction