missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-310.2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110698
This item is not returnable.
View return policy
Description
MA1-10698 detects Streptococci Group A. MA1-10698 has been successfully used in ELISA applications. The MA1-10698 immunogen is UV-inactivated Group A Streptococcus cells.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
Specifications
| Streptococcus Group A | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| UV-inactivated Group A Streptococcus cells. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| HSA12-310.2 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction