missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus Group B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA17250
This item is not returnable.
View return policy
Description
PA1-7250 detects the group B Streptococcus. This antibody does not cross react with other Streptococcus groups. PA1-7250 has been successfully used in ELISA and immunofluorescence procedures. The PA1-7250 immunogen is Streptococci, group B.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
Specifications
| Streptococcus Group B | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Streptococci, group B. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction