missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus pneumoniae Antibody (128/390), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64569C
This item is not returnable.
View return policy
Description
Streptococcus pneumoniae Monoclonal specifically detects Streptococcus pneumoniae in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Streptococcus pneumoniae | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Bacteria | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 128/390 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Pneumococcal surface protein A, PSPA | |
| Native, NCTC 10319 | |
| 0.1 mL | |
| In a simple ELISA using Biogenesis Test Groups (BTGs) this is broadly reactive with S.pneumoniae. A small cross reactivity is observed with Haemophilus influenzae. Immunohistology: Reactive with isolates fixed by phenol, formalin and boiling. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction