missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus pneumoniae Recombinant Rabbit Monoclonal Antibody (0495)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA533308
This item is not returnable.
View return policy
Description
For ELISA, use MA5-33307 as conjugate antibody and MA5-33308 as capture antibody.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Specifications
| Streptococcus pneumoniae | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.5 | |
| Rabbit | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Lateral Flow, Immunocytochemistry | |
| 0495 | |
| Unconjugated | |
| Pneumococcus; s pneumoniae; S. pneumoniae | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction